Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013839.3 | NP_001013861.1 | 13, 14 | 1587 | ||
NM_001145297.3 | NP_001138769.1 | 14, 15 | 1740 | |||
NM_001145298.3 | NP_001138770.1 | 13, 14 | 1563 | |||
NM_001145299.3 | NP_001138771.1 | 14, 15 | 1656 | |||
NM_001282313.1 | NP_001269242.1 | 13, 14 | 1616 | |||
NM_015219.4 | NP_056034.2 | 12, 13 | 1494 | |||
XM_006721786.3 | XP_006721849.1 | 15, 16 | 1694 | |||
XM_006721787.3 | XP_006721850.1 | 14, 15 | 2060 | |||
XM_006721788.3 | XP_006721851.1 | 14, 15 | 1601 | |||
XM_006721789.3 | XP_006721852.1 | 13, 14 | 1532 | |||
XM_011524571.2 | XP_011522873.1 | 14, 15 | 1971 | |||
XM_017024397.1 | XP_016879886.1 | 10, 11 | 1096 | |||
GenBank mRNA | AB028990.1 | - | 13, 14 | 1510 | ||
AK022552.1 | - | 4, 5 | 278 | |||
AK023832.1 | - | 12, 13 | 1402 | |||
AK098284.1 | - | 4, 5 | 1759 | |||
AK295871.1 | - | 13, 14 | 1568 | |||
AL834324.1 | - | 13, 14 | 1558 | |||
BC011045.1 | - | 13, 14 | 1521 | 4215228 | ||
BC018466.1 | - | 14, 15 | 1648 | 3867492 | ||
BX647763.1 | - | 11, 12 | 1255 | |||
FJ457119.1 | - | 13, 14 | 1468 | |||
FJ457120.1 | - | 14, 15 | 1561 |