Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260491.1 | NP_001247420.1 | 10 | 1127 | ||
NM_015266.2 | NP_056081.1 | 10 | 1079 | |||
NR_048537.1 | 9 | 1044 | ||||
NR_048538.1 | 8 | 960 | ||||
NR_048539.1 | 8 | 916 | ||||
NR_048540.1 | 8 | 942 | ||||
XM_006723756.1 | XP_006723819.1 | 11 | 1040 | |||
XM_011528736.1 | XP_011527038.1 | 11 | 1088 | |||
XM_011528737.1 | XP_011527039.1 | 10 | 961 | |||
XM_011528738.1 | XP_011527040.1 | 11 | 1088 | |||
XM_011528739.1 | XP_011527041.1 | 10 | 965 | |||
XM_011528740.1 | XP_011527042.1 | 11 | 1088 | |||
XM_011528741.1 | XP_011527043.1 | 10 | 989 | |||
XM_011528742.1 | XP_011527044.1 | 9 | 861 | |||
XM_011528743.2 | XP_011527045.1 | 3 | 225 | |||
XM_011528744.2 | XP_011527046.1 | 2 | 56 | |||
XM_017027754.1 | XP_016883243.1 | 11 | 1088 | |||
XM_017027755.1 | XP_016883244.1 | 10 | 949 | |||
XM_017027756.1 | XP_016883245.1 | 11 | 1037 | |||
GenBank mRNA | AB023156.2 | - | 10 | 911 | ||
AK295665.1 | - | 8 | 762 | |||
AK295803.1 | - | 9 | 719 | |||
AK295822.1 | - | 9 | 837 | |||
AK300318.1 | - | 10 | 1012 | |||
AK308328.1 | - | 8 | 746 | |||
AK310530.1 | - | 10 | 881 | |||
BC112213.1 | - | 10 | 901 | 8327681 | ||
BX430866.2 | - | 5 | 405 | |||
CR749638.1 | - | 10 | 956 |