Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KANSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193465.1 | NP_001180394.1 | 16 | 5037 | ||
NM_001193466.1 | NP_001180395.1 | 15 | 5254 | |||
NM_015443.3 | NP_056258.1 | 15 | 4956 | |||
XM_006721823.1 | XP_006721886.1 | 16 | 5040 | |||
XM_006721824.3 | XP_006721887.1 | 16 | 5224 | |||
XM_011524628.2 | XP_011522930.1 | 16 | 5221 | |||
XM_011524629.2 | XP_011522931.1 | 15 | 5122 | |||
XM_011524630.2 | XP_011522932.1 | 15 | 5035 | |||
XM_011524631.2 | XP_011522933.1 | 15 | 5033 | |||
XM_011524632.2 | XP_011522934.1 | 15 | 3650 | |||
GenBank mRNA | AK094946.1 | - | 7 | 2762 | ||
AL117476.1 | - | 4 | 2126 | |||
AL137317.1 | - | 11 | 3198 | |||
BC035892.1 | - | 10 | 3063 | |||
BC098376.1 | - | 15 | 4959 | 6181132 | ||
BX538006.1 | - | 15 | 5259 | |||
BX648760.1 | - | 8 | 9011 |