Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114093.1 | NP_001107565.1 | 11 | 3187 | ||
NM_015578.2 | NP_056393.2 | 10 | 3130 | |||
XM_017026575.1 | XP_016882064.1 | 9 | 2972 | |||
XM_017026576.1 | XP_016882065.1 | 10 | 3029 | |||
XM_017026577.1 | XP_016882066.1 | 9 | 2966 | |||
GenBank mRNA | AK027643.1 | - | 10 | 3016 | ||
AK127401.1 | - | 7 | 3276 | |||
AL833221.1 | - | 8 | 2621 |