Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016013.3 | NP_057097.2 | 3 | 1104 | ||
NR_045620.1 | 3 | 1104 | ||||
XM_006720555.2 | XP_006720618.1 | 3 | 1034 | |||
XM_011521658.1 | XP_011519960.1 | 3 | 968 | |||
XM_011521659.2 | XP_011519961.1 | 3 | 1131 | |||
XM_017022293.1 | XP_016877782.1 | 3 | 1035 | |||
GenBank mRNA | AF151823.1 | - | 3 | 1067 | ||
AK309889.1 | - | 3 | 1104 | |||
AK310983.1 | - | 3 | 1104 | |||
BC000780.2 | - | 3 | 1088 | 3504355 | ||
BG192055.1 | - | 1 | 136 |