Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308080.1 | NP_001295009.1 | 5 | 532 | ||
NM_016144.3 | NP_057228.1 | 5 | 505 | |||
XM_011543458.2 | XP_011541760.1 | 5 | 889 | |||
XM_011543460.2 | XP_011541762.1 | 5 | 888 | |||
XM_017009553.1 | XP_016865042.1 | 5 | 627 | |||
XM_017009555.1 | XP_016865044.1 | 5 | 887 | |||
XM_017009556.1 | XP_016865045.1 | 5 | 887 | |||
GenBank mRNA | AF078857.1 | - | 5 | 460 | ||
AF161423.1 | - | 5 | 456 | |||
AK002147.1 | - | 5 | 481 | |||
AY542165.1 | - | 5 | 428 | |||
BC005179.1 | - | 5 | 451 | 3683093 | ||
BI859871.1 | - | 5 | 521 | |||
DA001572.1 | - | 5 | 532 |