Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016520.2 | NP_057604.1 | 1, 2 | 158 | ||
GenBank mRNA | AF151054.1 | - | 1, 2 | 147 | ||
AF218421.1 | - | 1, 2 | 113 | |||
AK057004.1 | - | 1, 2 | 140 | |||
BC007664.2 | - | 1, 2 | 111 | 3140172 | ||
BC017570.1 | - | 1, 2 | 115 | 4109447 | ||
CB128496.1 | - | 1, 2 | 158 |