Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPHOSPH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017520.3 | NP_059990.2 | 13, 14 | 2650 | ||
XM_011535124.2 | XP_011533426.1 | 14, 15 | 2776 | |||
GenBank mRNA | AK056785.1 | - | 6, 7 | 697 | ||
BC003542.1 | - | 13, 14 | 2558 | 3611719 | ||
BC046214.1 | - | 7, 8 | 837 |