Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF562 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130031.1 | NP_001123503.1 | 6 | 5216 | ||
NM_001130032.1 | NP_001123504.1 | 6 | 5163 | |||
NM_001300885.1 | NP_001287814.1 | 6 | 5213 | |||
NM_017656.3 | NP_060126.2 | 4 | 5000 | |||
XM_005259941.3 | XP_005259998.1 | 6 | 5583 | |||
XM_005259944.3 | XP_005260001.1 | 5 | 5319 | |||
XM_005259951.3 | XP_005260008.1 | 5 | 5278 | |||
XM_005259952.3 | XP_005260009.1 | 5 | 5300 | |||
XM_011528100.2 | XP_011526402.1 | 4 | 5162 | |||
XM_011528102.2 | XP_011526404.1 | 3 | 5086 | |||
XM_017026898.1 | XP_016882387.1 | 6 | 5580 | |||
XM_017026899.1 | XP_016882388.1 | 5 | 5352 | |||
GenBank mRNA | AK000086.1 | - | 4 | 4957 |