Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PQLC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040125.1 | NP_001035214.1 | 3, 4 | 1026 | ||
NM_001040126.1 | NP_001035215.1 | 3, 4 | 414 | |||
NM_017765.2 | NP_060235.2 | 4, 5 | 656 | |||
NR_109848.1 | 4, 5 | 656 | ||||
XM_005245916.2 | XP_005245973.1 | 3, 4 | 439 | |||
XM_011541634.2 | XP_011539936.1 | 4, 5 | 608 | |||
GenBank mRNA | AK000327.1 | - | 4, 5 | 577 | ||
AK125580.1 | - | 3, 4 | 944 | |||
AK130171.1 | - | 4, 5 | 574 | |||
AK130175.1 | - | 4, 5 | 576 | |||
BC015324.1 | - | 3, 4 | 319 | 4426038 | ||
KU178659.1 | - | 1, 2 | 109 |