Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC41A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008485.1 | NP_001008485.1 | 9 | 1314 | ||
NM_001008486.1 | NP_001008486.1 | 8 | 1206 | |||
NM_001008487.1 | NP_001008487.1 | 8 | 1233 | |||
NM_001164475.1 | NP_001157947.1 | 8 | 935 | |||
NM_017836.3 | NP_060306.3 | 9 | 1314 | |||
XM_005247559.1 | XP_005247616.1 | 9 | 1192 | |||
XM_005247562.1 | XP_005247619.1 | 9 | 1208 | |||
XM_005247563.2 | XP_005247620.1 | 9 | 1237 | |||
XM_005247564.1 | XP_005247621.1 | 9 | 1192 | |||
XM_005247565.2 | XP_005247622.1 | 8 | 1296 | |||
XM_006713681.2 | XP_006713744.1 | 9 | 1154 | |||
XM_011512945.1 | XP_011511247.1 | 8 | 1341 | |||
XM_017006705.1 | XP_016862194.1 | 9 | 1181 | |||
XM_017006706.1 | XP_016862195.1 | 9 | 1208 | |||
XM_017006707.1 | XP_016862196.1 | 9 | 1231 | |||
XM_017006708.1 | XP_016862197.1 | 8 | 1039 | |||
XM_017006709.1 | XP_016862198.1 | 8 | 1094 | |||
GenBank mRNA | AK000480.1 | - | 8 | 1037 | ||
AK021925.1 | - | 9 | 1000 | |||
AK022780.1 | - | 8 | 1035 | |||
AK091671.1 | - | 10 | 1118 | |||
AK093380.1 | - | 8 | 1230 | |||
AK094051.1 | - | 4 | 1323 | |||
AK298070.1 | - | 8 | 944 | |||
AW938624.1 | - | 2 | 190 | |||
BC009039.2 | - | 9 | 1147 | 3852588 | ||
BC009444.1 | - | 9 | 1132 | |||
BC028241.1 | - | 8 | 1130 | 5243239 |