Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF532 SIRNA Assays ›
Gene Symbol:
ZNF532
Gene Name:
zinc finger protein 532
Gene Aliases:
-
Chromosome Location:
Chr.18: 58862600 - 58986480 on Build GRCh38
UniGene ID:
Hs.529023
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
147699
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF532
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318726.1
NP_001305655.1
3
784
NM_001318727.1
NP_001305656.1
3
342
NM_001318728.1
NP_001305657.1
4
444
NM_018181.5
NP_060651.2
4
866
XM_011526054.2
XP_011524356.1
3
591
XM_011526067.2
XP_011524369.1
1
107
XM_017025809.1
XP_016881298.1
1
107
XM_017025810.1
XP_016881299.1
6
1124
XM_017025811.1
XP_016881300.1
4
1187
XM_017025812.1
XP_016881301.1
4
424
XM_017025813.1
XP_016881302.1
3
383
XM_017025814.1
XP_016881303.1
3
338
XM_017025815.1
XP_016881304.1
2
674
XM_017025816.1
XP_016881305.1
4
471
XM_017025817.1
XP_016881306.1
4
672
XM_017025818.1
XP_016881307.1
3
442
XM_017025819.1
XP_016881308.1
4
495
XM_017025820.1
XP_016881309.1
3
591
XM_017025821.1
XP_016881310.1
3
784
XM_017025822.1
XP_016881311.1
3
592
XM_017025823.1
XP_016881312.1
2
349
XM_017025824.1
XP_016881313.1
2
351
XM_017025825.1
XP_016881314.1
3
991
XM_017025826.1
XP_016881315.1
3
592
XM_017025827.1
XP_016881316.1
3
413
XM_017025828.1
XP_016881317.1
3
592
XM_017025829.1
XP_016881318.1
2
350
XM_017025830.1
XP_016881319.1
2
352
XM_017025831.1
XP_016881320.1
3
592
GenBank mRNA
AB046849.2
-
3
302
AF295077.1
-
4
651
AK128058.1
-
4
868
AY039256.1
-
3
979
BC036366.1
-
4
354
4828639
BC130618.1
-
3
436
40146649
BC130620.1
-
4
518
40146651
DA108216.1
-
4
424
DA191371.1
-
4
444
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0