Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF532 SIRNA Assays ›
Gene Symbol:
ZNF532
Gene Name:
zinc finger protein 532
Gene Aliases:
-
Chromosome Location:
Chr.18: 58862600 - 58986480 on Build GRCh38
UniGene ID:
Hs.529023
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
147700
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF532
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318726.1
NP_001305655.1
3
1056
NM_001318727.1
NP_001305656.1
3
614
NM_001318728.1
NP_001305657.1
4
716
NM_018181.5
NP_060651.2
4
1138
XM_011526054.2
XP_011524356.1
3
863
XM_011526067.2
XP_011524369.1
1
379
XM_017025809.1
XP_016881298.1
1
379
XM_017025810.1
XP_016881299.1
6
1396
XM_017025811.1
XP_016881300.1
4
1459
XM_017025812.1
XP_016881301.1
4
696
XM_017025813.1
XP_016881302.1
3
655
XM_017025814.1
XP_016881303.1
3
610
XM_017025815.1
XP_016881304.1
2
946
XM_017025816.1
XP_016881305.1
4
743
XM_017025817.1
XP_016881306.1
4
944
XM_017025818.1
XP_016881307.1
3
714
XM_017025819.1
XP_016881308.1
4
767
XM_017025820.1
XP_016881309.1
3
863
XM_017025821.1
XP_016881310.1
3
1056
XM_017025822.1
XP_016881311.1
3
864
XM_017025823.1
XP_016881312.1
2
621
XM_017025824.1
XP_016881313.1
2
623
XM_017025825.1
XP_016881314.1
3
1263
XM_017025826.1
XP_016881315.1
3
864
XM_017025827.1
XP_016881316.1
3
685
XM_017025828.1
XP_016881317.1
3
864
XM_017025829.1
XP_016881318.1
2
622
XM_017025830.1
XP_016881319.1
2
624
XM_017025831.1
XP_016881320.1
3
864
GenBank mRNA
AB046849.2
-
3
574
AF295077.1
-
4
923
AK128058.1
-
4
1140
AY039256.1
-
3
1251
BC036366.1
-
4
626
4828639
BC130618.1
-
3
708
40146649
BC130620.1
-
4
790
40146651
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0