Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286772.1 | NP_001273701.1 | 10 | 3271 | ||
NM_018201.4 | NP_060671.3 | 12 | 3646 | |||
XM_005252060.2 | XP_005252117.1 | 11 | 3549 | |||
GenBank mRNA | AK001605.1 | - | 10 | 3135 | ||
AK027457.1 | - | 4 | 2694 | |||
AK123828.1 | - | 8 | 3262 | |||
BC014898.1 | - | 12 | 3534 | |||
BC150311.1 | - | 12 | 3559 | 8860447 |