Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307943.1 | NP_001294872.1 | 2 | 298 | ||
NM_018259.5 | NP_060729.2 | 2 | 298 | |||
XM_005253016.1 | XP_005253073.1 | 2 | 298 | |||
XM_011520218.2 | XP_011518520.1 | 2 | 298 | |||
XM_017018010.1 | XP_016873499.1 | 2 | 298 | |||
GenBank mRNA | AK001752.1 | - | 2 | 251 | ||
BC017238.2 | - | 2 | 247 | 3940012 | ||
BC033000.1 | - | 2 | 244 | 4830377 | ||
DA106610.1 | - | 2 | 298 |