Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CASC1 SIRNA Assays ›
Gene Symbol:
CASC1
Gene Name:
cancer susceptibility candidate 1
Gene Aliases:
LAS1, PPP1R54
Chromosome Location:
Chr.12: 25107057 - 25195177 on Build GRCh38
UniGene ID:
Hs.407771
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
147714
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CASC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001082972.2
NP_001076441.1
8
871
NM_001082973.2
NP_001076442.1
7
663
NM_001204101.2
NP_001191030.1
5
486
NM_001204102.2
NP_001191031.1
9
719
NM_001319977.1
NP_001306906.1
6
571
NM_001319978.1
NP_001306907.1
7
837
NM_018272.4
NP_060742.3
8
681
XM_005253406.1
XP_005253463.1
8
795
XM_005253408.1
XP_005253465.1
7
557
XM_005253409.1
XP_005253466.1
6
571
XM_006719101.1
XP_006719164.1
8
795
XM_011520723.1
XP_011519025.1
8
795
XM_011520726.1
XP_011519028.1
8
795
XM_011520727.1
XP_011519029.1
8
795
XM_017019560.1
XP_016875049.1
7
760
XM_017019561.1
XP_016875050.1
6
767
XM_017019562.1
XP_016875051.1
7
645
XM_017019563.1
XP_016875052.1
8
677
XM_017019564.1
XP_016875053.1
7
645
XM_017019565.1
XP_016875054.1
8
795
XM_017019566.1
XP_016875055.1
8
1006
XM_017019567.1
XP_016875056.1
5
450
XM_017019568.1
XP_016875057.1
6
589
GenBank mRNA
AK001783.1
-
2
86
AK093102.1
-
8
649
AK297998.1
-
5
450
AK302235.1
-
7
627
AK304602.1
-
6
571
AY423543.1
-
6
579
BC028415.1
-
8
812
4823620
BC031047.1
-
8
662
BC047415.1
-
9
717
5269578
BC057795.1
-
7
661
5266518
BC117281.1
-
5
496
BC117283.1
-
5
496
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-jlg4l:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline