Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322019.1 | NP_001308948.1 | 3, 4 | 569 | ||
NM_001322020.1 | NP_001308949.1 | 3, 4 | 569 | |||
NM_004544.3 | NP_004535.1 | 3, 4 | 569 | |||
NR_136155.1 | 3, 4 | 569 | ||||
NR_136156.1 | 3, 4 | 569 | ||||
NR_136157.1 | 2, 3 | 400 | ||||
NR_136158.1 | 3, 4 | 569 | ||||
XM_011511228.2 | XP_011509530.1 | 3, 4 | 556 | |||
GenBank mRNA | AB209440.1 | - | 3, 4 | 490 | ||
AF087661.1 | - | 3, 4 | 489 | |||
AF087854.1 | - | 3, 4 | 508 | |||
AF453834.1 | - | 3, 4 | 468 | |||
AK290778.1 | - | 3, 4 | 491 | |||
BC003417.1 | - | 3, 4 | 503 | 3451514 | ||
BC031332.1 | - | 3, 4 | 474 | 4872196 |