Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020936.2 | NP_065987.1 | 4, 5 | 418 | ||
NR_135104.1 | 4, 5 | 418 | ||||
NR_135105.1 | 4, 5 | 418 | ||||
XM_011538608.2 | XP_011536910.1 | 4, 5 | 406 | |||
XM_017019710.1 | XP_016875199.1 | 4, 5 | 403 | |||
XM_017019711.1 | XP_016875200.1 | 3, 4 | 458 | |||
XM_017019712.1 | XP_016875201.1 | 4, 5 | 256 | |||
XM_017019713.1 | XP_016875202.1 | 4, 5 | 433 | |||
XM_017019714.1 | XP_016875203.1 | 5, 6 | 591 | |||
XM_017019715.1 | XP_016875204.1 | 4, 5 | 404 | |||
XM_017019716.1 | XP_016875205.1 | 4, 5 | 403 | |||
XM_017019717.1 | XP_016875206.1 | 4, 5 | 365 | |||
XM_017019718.1 | XP_016875207.1 | 2, 3 | 783 | |||
XM_017019719.1 | XP_016875208.1 | 4, 5 | 374 | |||
XM_017019721.1 | XP_016875210.1 | 4, 5 | 404 | |||
GenBank mRNA | AK074219.1 | - | 4, 5 | 370 | ||
AK301043.1 | - | 4, 5 | 381 | |||
AK307384.1 | - | 4, 5 | 394 | |||
BC030020.2 | - | 4, 5 | 362 | 4821394 |