Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320635.1 | NP_001307564.1 | 5 | 1135 | ||
NM_001320636.1 | NP_001307565.1 | 6 | 985 | |||
NM_020980.4 | NP_066190.2 | 6 | 1353 | |||
GenBank mRNA | AB008775.1 | - | 6 | 1199 | ||
AF016495.1 | - | 6 | 1211 | |||
AK300055.1 | - | 6 | 958 | |||
BC026258.1 | - | 6 | 1168 | 4734559 |