Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021203.3 | NP_067026.3 | 8 | 1551 | ||
GenBank mRNA | AF141882.3 | - | 7 | 1282 | ||
AK027525.1 | - | 7 | 1282 | |||
AK075531.1 | - | 7 | 1320 | |||
BC063001.1 | - | 7 | 1295 | 5185997 | ||
BC065299.1 | - | 7 | 1264 | 6146939 |