Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105539.2 | NP_001099009.1 | 2 | 1805 | ||
NM_001277145.1 | NP_001264074.1 | 2 | 496 | |||
NM_023929.4 | NP_076418.3 | 2 | 1805 | |||
GenBank mRNA | AJ319673.1 | - | 2 | 1805 | ||
AK056913.1 | - | 2 | 496 | |||
BC050061.1 | - | 1 | 106 | |||
BC127097.1 | - | 3 | 1087 | |||
BC127098.1 | - | 3 | 817 | 40124012 | ||
BC148254.1 | - | 3 | 775 |