Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105539.2 | NP_001099009.1 | 6 | 3260 | ||
NM_001277145.1 | NP_001264074.1 | 6 | 1951 | |||
NM_023929.4 | NP_076418.3 | 7 | 3188 | |||
GenBank mRNA | AJ319673.1 | - | 7 | 3188 | ||
AK022814.1 | - | 6 | 837 | |||
AK056913.1 | - | 6 | 1951 | |||
BC050061.1 | - | 5 | 1561 | |||
BC127097.1 | - | 7 | 2542 | |||
BC127098.1 | - | 7 | 2272 | 40124012 | ||
BC148254.1 | - | 7 | 2224 |