Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011667.2 | NP_001011667.1 | 5 | 899 | ||
NM_001011668.2 | NP_001011668.1 | 5 | 855 | |||
NM_001011669.2 | NP_001011669.1 | 5 | 879 | |||
NM_001011670.2 | NP_001011670.1 | 4 | 751 | |||
NM_001011671.2 | NP_001011671.1 | 4 | 707 | |||
NM_001317858.1 | NP_001304787.1 | 5 | 854 | |||
NM_001317859.1 | NP_001304788.1 | 4 | 682 | |||
NM_024300.4 | NP_077276.2 | 5 | 835 | |||
NR_133934.1 | 6 | 974 | ||||
NR_133935.1 | 6 | 949 | ||||
NR_133936.1 | 6 | 930 | ||||
NR_133937.1 | 4 | 747 | ||||
GenBank mRNA | AK095922.1 | - | 4 | 556 | ||
AK225982.1 | - | 5 | 694 | |||
AY070434.1 | - | 5 | 727 | |||
BC002546.2 | - | 5 | 674 | 3139026 | ||
BI911444.1 | - | 5 | 685 | |||
BM695222.1 | - | 4 | 591 | |||
CN256918.1 | - | 4 | 512 |