Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011667.2 | NP_001011667.1 | 5 | 916 | ||
NM_001011668.2 | NP_001011668.1 | 5 | 872 | |||
NM_001011669.2 | NP_001011669.1 | 5 | 896 | |||
NM_001011670.2 | NP_001011670.1 | 4 | 768 | |||
NM_001011671.2 | NP_001011671.1 | 4 | 724 | |||
NM_001317858.1 | NP_001304787.1 | 5 | 871 | |||
NM_001317859.1 | NP_001304788.1 | 4 | 699 | |||
NM_024300.4 | NP_077276.2 | 5 | 852 | |||
NR_133934.1 | 6 | 991 | ||||
NR_133935.1 | 6 | 966 | ||||
NR_133936.1 | 6 | 947 | ||||
NR_133937.1 | 4 | 764 | ||||
GenBank mRNA | AK095922.1 | - | 4 | 573 | ||
AK225982.1 | - | 5 | 711 | |||
AY070434.1 | - | 5 | 744 | |||
BC002546.2 | - | 5 | 691 | 3139026 | ||
BM695222.1 | - | 4 | 608 | |||
CN256918.1 | - | 4 | 529 |