Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011667.2 | NP_001011667.1 | 5 | 1143 | ||
NM_001011668.2 | NP_001011668.1 | 5 | 1099 | |||
NM_001011669.2 | NP_001011669.1 | 5 | 1123 | |||
NM_001011670.2 | NP_001011670.1 | 4 | 995 | |||
NM_001011671.2 | NP_001011671.1 | 4 | 951 | |||
NM_001317858.1 | NP_001304787.1 | 5 | 1098 | |||
NM_001317859.1 | NP_001304788.1 | 4 | 926 | |||
NM_024300.4 | NP_077276.2 | 5 | 1079 | |||
NR_133934.1 | 6 | 1218 | ||||
NR_133935.1 | 6 | 1193 | ||||
NR_133936.1 | 6 | 1174 | ||||
NR_133937.1 | 4 | 991 | ||||
GenBank mRNA | AK095922.1 | - | 4 | 800 | ||
AK225982.1 | - | 5 | 938 | |||
AY070434.1 | - | 5 | 971 | |||
BC002546.2 | - | 5 | 918 | 3139026 |