Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193528.1 | NP_001180457.1 | 4, 5 | 558 | ||
NM_024583.4 | NP_078859.2 | 4, 5 | 649 | |||
XM_005246853.2 | XP_005246910.1 | 4, 5 | 1054 | |||
XM_005246854.4 | XP_005246911.1 | 4, 5 | 2214 | |||
XM_005246855.2 | XP_005246912.1 | 4, 5 | 1102 | |||
XM_005246856.2 | XP_005246913.1 | 4, 5 | 1058 | |||
XM_005246857.2 | XP_005246914.1 | 4, 5 | 758 | |||
XM_011511839.2 | XP_011510141.1 | 3, 4 | 458 | |||
XM_017004909.1 | XP_016860398.1 | 4, 5 | 1646 | |||
XM_017004910.1 | XP_016860399.1 | 4, 5 | 1124 | |||
GenBank mRNA | AK057912.1 | - | 4, 5 | 627 | ||
AK295357.1 | - | 4, 5 | 542 | |||
AK296558.1 | - | 3, 4 | 448 | |||
BC031821.1 | - | 1, 2 | 110 | |||
BC119684.1 | - | 4, 5 | 600 | 40116568 | ||
BC119685.1 | - | 4, 5 | 600 | 40116570 | ||
BM725063.1 | - | 3, 4 | 640 |