Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193528.1 | NP_001180457.1 | 5 | 685 | ||
NM_024583.4 | NP_078859.2 | 5 | 776 | |||
XM_005246853.2 | XP_005246910.1 | 5 | 1181 | |||
XM_005246854.4 | XP_005246911.1 | 5 | 2341 | |||
XM_005246855.2 | XP_005246912.1 | 5 | 1229 | |||
XM_005246856.2 | XP_005246913.1 | 5 | 1185 | |||
XM_005246857.2 | XP_005246914.1 | 5 | 885 | |||
XM_011511839.2 | XP_011510141.1 | 4 | 585 | |||
XM_017004909.1 | XP_016860398.1 | 5 | 1773 | |||
XM_017004910.1 | XP_016860399.1 | 5 | 1251 | |||
GenBank mRNA | AK026795.1 | - | 1 | 123 | ||
AK057912.1 | - | 5 | 754 | |||
AK295357.1 | - | 5 | 669 | |||
AK296558.1 | - | 4 | 575 | |||
BC031821.1 | - | 2 | 237 | |||
BC119684.1 | - | 5 | 727 | 40116568 | ||
BC119685.1 | - | 5 | 727 | 40116570 |