Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC7 SIRNA Assays ›
Gene Symbol:
CCDC7
Gene Name:
coiled-coil domain containing 7
Gene Aliases:
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
Chromosome Location:
Chr.10: 32446082 - 32882864 on Build GRCh38
UniGene ID:
Hs.585464
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
147866
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321115.1
NP_001308044.1
26, 27
3169
XM_005252593.2
XP_005252650.1
7, 8
956
XM_006717505.1
XP_006717568.1
24, 25
2664
XM_006717506.1
XP_006717569.1
23, 24
2556
XM_011519663.1
XP_011517965.1
26, 27
2877
XM_011519664.1
XP_011517966.1
26, 27
2877
XM_011519666.1
XP_011517968.1
26, 27
2877
XM_011519667.1
XP_011517969.1
26, 27
2877
XM_011519668.1
XP_011517970.1
25, 26
2787
XM_011519669.1
XP_011517971.1
25, 26
2775
XM_011519670.1
XP_011517972.1
25, 26
2769
XM_011519671.1
XP_011517973.1
26, 27
2877
XM_011519672.1
XP_011517974.1
25, 26
2766
XM_011519673.1
XP_011517975.1
25, 26
2763
XM_011519674.1
XP_011517976.1
26, 27
2877
XM_011519675.1
XP_011517977.1
24, 25
2667
XM_011519676.1
XP_011517978.1
25, 26
2763
XM_011519677.1
XP_011517979.1
25, 26
2655
XM_011519678.1
XP_011517980.1
24, 25
2652
XM_011519679.1
XP_011517981.1
23, 24
2346
XM_011519680.2
XP_011517982.1
21, 22
2012
XM_011519683.2
XP_011517985.1
15, 16
1686
XM_011519684.2
XP_011517986.1
26, 27
2877
XM_011519685.1
XP_011517987.1
18, 19
1894
XM_011519686.2
XP_011517988.1
26, 27
2877
XM_011519687.1
XP_011517989.1
8, 9
801
XM_017016638.1
XP_016872127.1
26, 27
2877
XM_017016640.1
XP_016872129.1
26, 27
2877
XM_017016641.1
XP_016872130.1
21, 22
2306
XM_017016642.1
XP_016872131.1
21, 22
2379
XM_017016643.1
XP_016872132.1
21, 22
1996
XM_017016644.1
XP_016872133.1
26, 27
2877
XM_017016645.1
XP_016872134.1
20, 21
1920
XM_017016646.1
XP_016872135.1
17, 18
1807
XM_017016647.1
XP_016872136.1
26, 27
2877
XM_017016648.1
XP_016872137.1
13, 14
1528
XM_017016649.1
XP_016872138.1
13, 14
1468
XM_017016652.1
XP_016872141.1
10, 11
931
XM_017016653.1
XP_016872142.1
8, 9
841
GenBank mRNA
AK023093.1
-
7, 8
962
AK301824.1
-
4, 5
397
BC125090.1
-
10, 11
1131
40118176
BC125091.1
-
5, 6
487
40118177
DB026046.1
-
3, 4
315
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-pfczg:80/100.66.78.229:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline