Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GREB1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142966.1 | NP_001136438.1 | 4 | 480 | ||
XM_006722547.2 | XP_006722610.1 | 3 | 314 | |||
XM_011526179.2 | XP_011524481.1 | 3 | 314 | |||
XM_017025988.1 | XP_016881477.1 | 3 | 314 | |||
XM_017025989.1 | XP_016881478.1 | 3 | 387 | |||
XM_017025990.1 | XP_016881479.1 | 3 | 314 | |||
XM_017025991.1 | XP_016881480.1 | 3 | 314 | |||
XM_017025992.1 | XP_016881481.1 | 3 | 314 | |||
XM_017025993.1 | XP_016881482.1 | 3 | 387 | |||
XM_017025994.1 | XP_016881483.1 | 3 | 314 | |||
XM_017025995.1 | XP_016881484.1 | 3 | 314 | |||
XM_017025996.1 | XP_016881485.1 | 3 | 314 | |||
GenBank mRNA | AB051559.1 | - | 4 | 480 | ||
AK309723.1 | - | 3 | 314 | |||
BC125017.1 | - | 4 | 364 |