Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GREB1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142966.1 | NP_001136438.1 | 25 | 4571 | ||
XM_006722547.2 | XP_006722610.1 | 25 | 4534 | |||
XM_011526179.2 | XP_011524481.1 | 25 | 4534 | |||
XM_017025988.1 | XP_016881477.1 | 25 | 4534 | |||
XM_017025989.1 | XP_016881478.1 | 25 | 4607 | |||
XM_017025990.1 | XP_016881479.1 | 25 | 4534 | |||
XM_017025991.1 | XP_016881480.1 | 25 | 4450 | |||
XM_017025992.1 | XP_016881481.1 | 24 | 4405 | |||
XM_017025993.1 | XP_016881482.1 | 24 | 4478 | |||
XM_017025994.1 | XP_016881483.1 | 24 | 4207 | |||
XM_017025995.1 | XP_016881484.1 | 24 | 4123 | |||
XM_017025996.1 | XP_016881485.1 | 23 | 4078 | |||
GenBank mRNA | AK023749.1 | - | 12 | 2324 | ||
AK293321.1 | - | 13 | 2518 |