Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM214B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317991.1 | NP_001304920.1 | 9 | 2043 | ||
NM_025182.3 | NP_079458.2 | 9 | 2044 | |||
NR_134455.1 | 10 | 1680 | ||||
XM_005251588.1 | XP_005251645.1 | 9 | 2178 | |||
XM_005251590.1 | XP_005251647.1 | 9 | 1981 | |||
XM_005251591.1 | XP_005251648.1 | 9 | 2017 | |||
XM_005251592.1 | XP_005251649.1 | 9 | 1458 | |||
XM_005251593.1 | XP_005251650.1 | 9 | 1340 | |||
XM_005251594.1 | XP_005251651.1 | 9 | 1214 | |||
XM_005251596.4 | XP_005251653.1 | 9 | 1202 | |||
XM_005251597.4 | XP_005251654.1 | 9 | 1245 | |||
XM_005251598.4 | XP_005251655.1 | 9 | 1180 | |||
XM_011518037.1 | XP_011516339.1 | 9 | 2000 | |||
XM_011518039.1 | XP_011516341.1 | 8 | 2109 | |||
XM_011518040.1 | XP_011516342.1 | 10 | 2064 | |||
XM_011518043.1 | XP_011516345.1 | 10 | 1482 | |||
XM_011518044.1 | XP_011516346.1 | 10 | 1402 | |||
XM_017015169.1 | XP_016870658.1 | 9 | 2314 | |||
XM_017015170.1 | XP_016870659.1 | 10 | 1698 | |||
GenBank mRNA | AB040972.1 | - | 8 | 4774 | ||
AK022837.1 | - | 10 | 1558 | |||
AL583909.1 | - | 9 | 2043 | |||
BC004406.1 | - | 9 | 2029 | 3638496 |