Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM214B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317991.1 | NP_001304920.1 | 9 | 2563 | ||
NM_025182.3 | NP_079458.2 | 9 | 2564 | |||
NR_134455.1 | 10 | 2200 | ||||
XM_005251588.1 | XP_005251645.1 | 9 | 2698 | |||
XM_005251590.1 | XP_005251647.1 | 9 | 2501 | |||
XM_005251591.1 | XP_005251648.1 | 9 | 2537 | |||
XM_005251592.1 | XP_005251649.1 | 9 | 1978 | |||
XM_005251593.1 | XP_005251650.1 | 9 | 1860 | |||
XM_005251594.1 | XP_005251651.1 | 9 | 1734 | |||
XM_005251596.4 | XP_005251653.1 | 9 | 1722 | |||
XM_005251597.4 | XP_005251654.1 | 9 | 1765 | |||
XM_005251598.4 | XP_005251655.1 | 9 | 1700 | |||
XM_011518037.1 | XP_011516339.1 | 9 | 2520 | |||
XM_011518039.1 | XP_011516341.1 | 8 | 2629 | |||
XM_011518040.1 | XP_011516342.1 | 10 | 2584 | |||
XM_011518043.1 | XP_011516345.1 | 10 | 2002 | |||
XM_011518044.1 | XP_011516346.1 | 10 | 1922 | |||
XM_017015169.1 | XP_016870658.1 | 9 | 2834 | |||
XM_017015170.1 | XP_016870659.1 | 10 | 2218 | |||
GenBank mRNA | AB040972.1 | - | 8 | 5294 | ||
AK021622.1 | - | 8 | 1778 | |||
AK022837.1 | - | 10 | 2078 | |||
AL583909.1 | - | 9 | 2563 | |||
BC004406.1 | - | 9 | 2549 | 3638496 |