Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADPGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031284.4 | NP_112574.3 | 5, 6 | 942 | ||
NR_023318.1 | 5, 6 | 1155 | ||||
NR_023319.1 | 3, 4 | 653 | ||||
XM_006720712.3 | XP_006720775.1 | 5, 6 | 942 | |||
XM_006720713.3 | XP_006720776.1 | 5, 6 | 1103 | |||
XM_011522104.2 | XP_011520406.1 | 5, 6 | 942 | |||
XM_017022664.1 | XP_016878153.1 | 1, 2 | 883 | |||
GenBank mRNA | AF204270.1 | - | 4, 5 | 531 | ||
AK055526.1 | - | 3, 4 | 644 | |||
AK075560.1 | - | 5, 6 | 877 | |||
AK293282.1 | - | 5, 6 | 634 | |||
AK294391.1 | - | 4, 5 | 679 | |||
BC006112.1 | - | 5, 6 | 884 | 3347312 | ||
BC044589.1 | - | 5, 6 | 1101 |