Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029835.2 | NP_001025006.1 | 2 | 321 | ||
NM_001167935.1 | NP_001161407.1 | 2 | 333 | |||
NM_031443.3 | NP_113631.1 | 2 | 333 | |||
NR_030770.1 | 2 | 269 | ||||
XM_006715786.3 | XP_006715849.1 | 2 | 929 | |||
XM_011515561.2 | XP_011513863.1 | 2 | 283 | |||
XM_011515562.1 | XP_011513864.1 | 2 | 314 | |||
XM_017012671.1 | XP_016868160.1 | 2 | 283 | |||
GenBank mRNA | AF370392.1 | - | 2 | 333 | ||
AK025807.1 | - | 1 | 48 | |||
AK098005.1 | - | 2 | 929 | |||
AK298671.1 | - | 2 | 283 | |||
AK303534.1 | - | 2 | 285 | |||
BC004903.2 | - | 2 | 268 | 3501896 | ||
BC008859.2 | - | 2 | 268 | 4124185 | ||
BC016832.2 | - | 2 | 262 | 2924210 | ||
BC063663.1 | - | 2 | 268 | |||
DA420491.1 | - | 2 | 257 |