Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029835.2 | NP_001025006.1 | 6 | 796 | ||
NM_001167934.1 | NP_001161406.1 | 5 | 634 | |||
NM_031443.3 | NP_113631.1 | 6 | 808 | |||
NR_030770.1 | 6 | 744 | ||||
XM_006715785.3 | XP_006715848.1 | 5 | 1686 | |||
XM_011515561.2 | XP_011513863.1 | 6 | 758 | |||
XM_011515562.1 | XP_011513864.1 | 6 | 789 | |||
XM_011515563.2 | XP_011513865.1 | 5 | 1686 | |||
XM_011515564.2 | XP_011513866.1 | 5 | 621 | |||
XM_017012671.1 | XP_016868160.1 | 6 | 758 | |||
XM_017012672.1 | XP_016868161.1 | 5 | 1686 | |||
XM_017012673.1 | XP_016868162.1 | 5 | 589 | |||
GenBank mRNA | AF370392.1 | - | 6 | 808 | ||
AK025807.1 | - | 5 | 523 | |||
AK098005.1 | - | 6 | 1404 | |||
AK124060.1 | - | 2 | 997 | |||
AK298671.1 | - | 4 | 537 | |||
AK303216.1 | - | 5 | 584 | |||
AK303534.1 | - | 4 | 492 | |||
BC004903.2 | - | 6 | 743 | 3501896 | ||
BC008859.2 | - | 6 | 743 | 4124185 | ||
BC016832.2 | - | 6 | 737 | 2924210 | ||
BC025958.1 | - | 2 | 140 | |||
BX641167.1 | - | 2 | 97 |