Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004375.4 | NP_004366.1 | 4 | 1929 | ||
NR_135677.1 | 4 | 1929 | ||||
XM_011524342.2 | XP_011522644.1 | 4 | 4168 | |||
XM_017024192.1 | XP_016879681.1 | 4 | 4168 | |||
XM_017024193.1 | XP_016879682.1 | 4 | 4168 | |||
GenBank mRNA | AF044321.1 | - | 4 | 1805 |
Gene Symbol: TOM1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW044673.1 | - | 1 | 210 | ||
AW139144.1 | - | 1 | 193 | |||
CB241257.1 | - | 1 | 194 |