Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AFAP1L2 SIRNA Assays ›
Gene Symbol:
AFAP1L2
Gene Name:
actin filament associated protein 1 like 2
Gene Aliases:
CTB-1144G6.4, KIAA1914, XB130
Chromosome Location:
Chr.10: 114280730 - 114405143 on Build GRCh38
UniGene ID:
Hs.501106
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
147943
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: AFAP1L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001936.2
NP_001001936.1
19
3402
NM_001287824.1
NP_001274753.1
20
3561
NM_032550.3
NP_115939.1
19
3390
XM_011540289.1
XP_011538591.1
19
3172
XM_011540290.1
XP_011538592.1
19
3160
XM_017016800.1
XP_016872289.1
20
3256
XM_017016801.1
XP_016872290.1
20
3253
XM_017016802.1
XP_016872291.1
20
3244
XM_017016803.1
XP_016872292.1
20
3241
XM_017016805.1
XP_016872294.1
20
3208
XM_017016806.1
XP_016872295.1
20
3205
XM_017016807.1
XP_016872296.1
19
3169
XM_017016808.1
XP_016872297.1
20
3196
XM_017016809.1
XP_016872298.1
20
3193
XM_017016810.1
XP_016872299.1
20
3350
XM_017016811.1
XP_016872300.1
20
3347
XM_017016812.1
XP_016872301.1
20
3338
XM_017016813.1
XP_016872302.1
20
3335
XM_017016816.1
XP_016872305.1
19
3123
XM_017016817.1
XP_016872306.1
19
3111
XM_017016819.1
XP_016872308.1
19
3007
XM_017016820.1
XP_016872309.1
21
3433
XM_017016821.1
XP_016872310.1
21
3430
XM_017016822.1
XP_016872311.1
19
2995
XM_017016823.1
XP_016872312.1
19
2959
XM_017016824.1
XP_016872313.1
19
2947
XM_017016825.1
XP_016872314.1
20
3349
XM_017016826.1
XP_016872315.1
20
3346
XM_017016827.1
XP_016872316.1
18
2877
XM_017016828.1
XP_016872317.1
18
2865
XM_017016830.1
XP_016872319.1
19
3112
XM_017016831.1
XP_016872320.1
18
3015
GenBank mRNA
AB067501.1
-
6
3110
AF474151.1
-
19
3128
AK123108.1
-
20
3160
BC024314.1
-
19
3368
3900921
BC033212.1
-
19
3109
4909140
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
inflammatory response
regulation of mitotic cell cycle
regulation of interleukin-6 production
positive regulation of interleukin-8 production
positive regulation of epidermal growth factor receptor signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of protein tyrosine kinase activity
SH3 domain binding
protein tyrosine kinase activator activity
signaling adaptor activity
SH2 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0