Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032853.3 | NP_116242.2 | 8 | 1436 | ||
NR_024247.1 | 8 | 1505 | ||||
XM_011528380.2 | XP_011526682.1 | 8 | 1426 | |||
XM_011528382.2 | XP_011526684.1 | 8 | 1426 | |||
XM_011528383.1 | XP_011526685.1 | 8 | 1426 | |||
XM_011528384.2 | XP_011526686.1 | 8 | 1426 | |||
XM_011528385.2 | XP_011526687.1 | 9 | 1627 | |||
XM_017027401.1 | XP_016882890.1 | 8 | 1426 | |||
GenBank mRNA | AK075241.1 | - | 2 | 66 | ||
AK309092.1 | - | 7 | 1324 | |||
AK310721.1 | - | 9 | 1539 | |||
BC110874.1 | - | 7 | 1402 | 4933735 | ||
BC130443.1 | - | 8 | 1465 | 40146474 | ||
BC144138.1 | - | 7 | 1352 | 9052658 | ||
BC144147.1 | - | 8 | 1465 | |||
BQ071619.1 | - | 4 | 467 |