Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286739.1 | NP_001273668.1 | 4, 5 | 492 | ||
NM_001286740.1 | NP_001273669.1 | 3, 4 | 357 | |||
NM_001286741.1 | NP_001273670.1 | 3, 4 | 847 | |||
NM_001286742.1 | NP_001273671.1 | 4, 5 | 589 | |||
NM_032907.4 | NP_116296.1 | 3, 4 | 478 | |||
NM_201265.1 | NP_957717.1 | 3, 4 | 407 | |||
XM_011522124.2 | XP_011520426.1 | 4, 5 | 694 | |||
GenBank mRNA | AY037166.1 | - | 3, 4 | 381 | ||
BC007913.2 | - | 3, 4 | 761 | 4302762 | ||
BC030055.1 | - | 4, 5 | 455 | 4873512 | ||
BC033919.1 | - | 4, 5 | 496 | 4904699 | ||
BG704085.1 | - | 3, 4 | 404 | |||
BI551787.1 | - | 3, 4 | 464 | |||
BP192909.1 | - | 3, 4 | 468 | |||
HY135530.1 | - | 3, 4 | 357 |