Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252678.1 | NP_001239607.1 | 7 | 511 | ||
NM_001320665.1 | NP_001307594.1 | 8 | 558 | |||
NM_033028.4 | NP_149017.2 | 8 | 558 | |||
NR_045565.1 | 9 | 616 | ||||
NR_045566.1 | 8 | 871 | ||||
XM_011521848.1 | XP_011520150.1 | 8 | 533 | |||
XM_011521849.1 | XP_011520151.1 | 7 | 421 | |||
XM_011521851.1 | XP_011520153.1 | 8 | 533 | |||
XM_017022450.1 | XP_016877939.1 | 8 | 839 | |||
XM_017022451.1 | XP_016877940.1 | 7 | 499 | |||
XM_017022452.1 | XP_016877941.1 | 7 | 729 | |||
XM_017022453.1 | XP_016877942.1 | 7 | 416 | |||
XM_017022454.1 | XP_016877943.1 | 6 | 366 | |||
GenBank mRNA | AF359281.1 | - | 8 | 530 | ||
AK075321.1 | - | 8 | 522 | |||
AK298922.1 | - | 9 | 588 | |||
AK302224.1 | - | 8 | 825 | |||
AK303706.1 | - | 7 | 506 | |||
AK310106.1 | - | 2 | 69 | |||
AK314507.1 | - | 8 | 530 | |||
AY457143.1 | - | 8 | 630 | |||
BC008923.2 | - | 5 | 335 | |||
BC027624.2 | - | 8 | 528 | 4824276 | ||
BE727117.1 | - | 7 | 499 | |||
BI562463.1 | - | 8 | 553 | |||
BX647855.1 | - | 7 | 482 | |||
DC372967.1 | - | 8 | 558 | |||
KU177936.1 | - | 8 | 509 |