Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC44A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286730.1 | NP_001273659.1 | 5, 6 | 760 | ||
NM_080546.4 | NP_536856.2 | 5, 6 | 760 | |||
XM_005251855.4 | XP_005251912.1 | 5, 6 | 765 | |||
XM_006717027.3 | XP_006717090.1 | 5, 6 | 573 | |||
XM_006717028.3 | XP_006717091.1 | 5, 6 | 573 | |||
XM_006717029.2 | XP_006717092.1 | 5, 6 | 573 | |||
XM_017014559.1 | XP_016870048.1 | 5, 6 | 748 | |||
XM_017014560.1 | XP_016870049.1 | 6, 7 | 738 | |||
GenBank mRNA | AJ272365.1 | - | 5, 6 | 713 | ||
AJ420812.1 | - | 5, 6 | 546 | |||
AK022549.1 | - | 1, 2 | 113 | |||
BC049203.1 | - | 5, 6 | 513 | 5492167 |