Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004398.3 | NP_004389.2 | 13 | 1856 | ||
GenBank mRNA | AB040537.1 | - | 13 | 1792 | ||
AB209884.1 | - | 13 | 1759 | |||
AK296752.1 | - | 11 | 1530 | |||
AK315216.1 | - | 13 | 1787 | |||
BC049217.1 | - | 13 | 1752 | |||
BC091521.1 | - | 13 | 1804 | 30388370 | ||
BC093654.1 | - | 13 | 1760 | 7939499 | ||
BC093656.1 | - | 13 | 1760 | 7939501 | ||
U28042.1 | - | 13 | 1792 |