Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOB3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130807.2 | NP_570719.1 | 5 | 1231 | ||
XM_011527683.2 | XP_011525985.1 | 5 | 1308 | |||
GenBank mRNA | AK094732.1 | - | 2 | 332 | ||
AK095471.1 | - | 5 | 1231 | |||
AL832474.1 | - | 5 | 1267 | |||
BC015049.1 | - | 4 | 1120 | 3919665 | ||
CR627409.1 | - | 5 | 854 |