Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYADM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001020818.2 | NP_001018654.1 | 2 | 2017 | ||
NM_001020819.2 | NP_001018655.1 | 3 | 1797 | |||
NM_001020820.2 | NP_001018656.1 | 3 | 1914 | |||
NM_001020821.2 | NP_001018657.1 | 3 | 1862 | |||
NM_001290188.1 | NP_001277117.1 | 3 | 1978 | |||
NM_001290189.1 | NP_001277118.1 | 3 | 2248 | |||
NM_001290190.1 | NP_001277119.1 | 3 | 1769 | |||
NM_001290191.1 | NP_001277120.1 | 3 | 1721 | |||
NM_001290192.1 | NP_001277121.1 | 3 | 1738 | |||
NM_001290193.1 | NP_001277122.1 | 3 | 1799 | |||
NM_001290194.1 | NP_001277123.1 | 2 | 1648 | |||
NM_138373.4 | NP_612382.1 | 3 | 1733 | |||
GenBank mRNA | AF087882.1 | - | 3 | 1845 | ||
AK027693.1 | - | 1 | 1353 | |||
AK075276.1 | - | 3 | 1716 | |||
AK093999.1 | - | 4 | 1659 | |||
AX775819.1 | - | 3 | 1712 | |||
AY358582.1 | - | 3 | 1697 | |||
BC095412.1 | - | 3 | 1714 | 30527914 |