Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138443.3 | NP_612452.1 | 8, 9 | 860 | ||
NR_026978.1 | 8, 9 | 900 | ||||
GenBank mRNA | AK024747.1 | - | 5, 6 | 323 | ||
AK097403.1 | - | 8, 9 | 841 | |||
AK315605.1 | - | 8, 9 | 806 | |||
AY360137.1 | - | 8, 9 | 860 | |||
BC005958.1 | - | 8, 9 | 836 | 4047707 | ||
BC014003.2 | - | 8, 9 | 833 | 3844806 |