Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICA1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288622.1 | NP_001275551.1 | 13 | 2983 | ||
NM_001288623.1 | NP_001275552.1 | 13 | 2928 | |||
NM_138468.5 | NP_612477.3 | 14 | 3110 | |||
GenBank mRNA | AK055978.1 | - | 13 | 2725 | ||
AK092310.1 | - | 13 | 2983 | |||
AL833904.1 | - | 14 | 2767 | |||
BC063876.1 | - | 13 | 2641 | 30377894 |