Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256129.1 | NP_001243058.1 | 6 | 699 | ||
NM_138795.3 | NP_620150.1 | 7 | 767 | |||
GenBank mRNA | AK127138.1 | - | 7 | 720 | ||
BC015408.1 | - | 7 | 698 | 4391821 | ||
BG876867.1 | - | 6 | 540 | |||
BM548962.1 | - | 7 | 752 |