Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195215.1 | NP_001182144.1 | 10, 11 | 1014 | ||
NM_001300858.1 | NP_001287787.1 | 10, 11 | 1165 | |||
NM_144977.4 | NP_659414.2 | 10, 11 | 1014 | |||
NR_125340.1 | 9, 10 | 954 | ||||
XM_005244931.2 | XP_005244988.1 | 9, 10 | 954 | |||
XM_006711192.2 | XP_006711255.1 | 11, 12 | 924 | |||
XM_006711193.3 | XP_006711256.1 | 11, 12 | 1056 | |||
XM_011509246.2 | XP_011507548.1 | 10, 11 | 875 | |||
XM_011509247.1 | XP_011507549.1 | 9, 10 | 815 | |||
XM_011509248.2 | XP_011507550.1 | 10, 11 | 808 | |||
XM_011509249.2 | XP_011507551.1 | 11, 12 | 1113 | |||
XM_011509251.2 | XP_011507553.1 | 10, 11 | 877 | |||
XM_017000470.1 | XP_016855959.1 | 11, 12 | 1002 | |||
XM_017000471.1 | XP_016855960.1 | 10, 11 | 864 | |||
XM_017000472.1 | XP_016855961.1 | 7, 8 | 534 | |||
GenBank mRNA | AK091207.1 | - | 9, 10 | 768 | ||
AL831839.1 | - | 4, 5 | 301 | |||
BC022561.1 | - | 10, 11 | 867 | 4793412 | ||
BC063877.1 | - | 10, 11 | 846 | 30377843 |