Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190233.1 | NP_001177162.1 | 6 | 805 | ||
NM_145261.3 | NP_660304.1 | 6 | 793 | |||
NR_033721.1 | 5 | 741 | ||||
NR_033722.1 | 5 | 713 | ||||
NR_033723.1 | 6 | 738 | ||||
NR_046073.1 | 3 | 587 | ||||
GenBank mRNA | BC073989.1 | - | 6 | 738 | 5760865 | |
BG492744.1 | - | 5 | 679 | |||
CB054787.1 | - | 3 | 509 |