Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTKN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145307.3 | NP_660350.2 | 6 | 829 | ||
XM_011539456.2 | XP_011537758.1 | 6 | 845 | |||
XM_011539457.2 | XP_011537759.1 | 6 | 845 | |||
XM_011539460.2 | XP_011537762.1 | 6 | 845 | |||
XM_017015842.1 | XP_016871331.1 | 5 | 747 | |||
XM_017015843.1 | XP_016871332.1 | 6 | 845 | |||
XM_017015844.1 | XP_016871333.1 | 6 | 845 | |||
GenBank mRNA | AY150309.1 | - | 6 | 673 |